Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004582.1 | NP_001004582.1 | 1 - 2 | 66 | 111 | |
GenBank mRNA | BC081638.1 | - | 1 - 2 | 66 | 111 | |
BC153573.1 | - | 1 - 2 | 66 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |