Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2k16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005963.2 | NP_001005963.2 | 5 - 6 | 918 | 130 | |
GenBank mRNA | BC083304.1 | - | 5 - 6 | 861 | 130 | |
BC165360.1 | - | 5 - 6 | 883 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |