Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008576.2 | NP_001008576.1 | 4 - 5 | 1003 | 75 | |
XM_017356481.2 | XP_017211970.1 | 4 - 5 | 855 | 75 | ||
XM_021476852.1 | XP_021332527.1 | 4 - 5 | 1050 | 75 | ||
XM_021476853.1 | XP_021332528.1 | 4 - 5 | 813 | 75 | ||
GenBank mRNA | BC086960.1 | - | 4 - 5 | 1003 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |