Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsf6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008732.1 | NP_001008732.1 | 4 - 5 | 629 | 91 | |
XM_009305734.2 | XP_009304009.1 | 3 - 4 | 91 | |||
XM_021479605.1 | XP_021335280.1 | 3 - 4 | 91 | |||
XM_021479606.1 | XP_021335281.1 | 4 - 5 | 91 | |||
GenBank mRNA | BC086820.1 | - | 4 - 5 | 629 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |