Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rybpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012373.2 | NP_001012373.1 | 3 - 4 | 504 | 59 | |
GenBank mRNA | AY885249.1 | - | 3 - 4 | 432 | 59 | |
BC125899.1 | - | 3 - 4 | 504 | 59 | ||
BC165507.1 | - | 3 - 4 | 466 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |