Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eef1akmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013327.1 | NP_001013345.1 | 4 - 5 | 401 | 119 | |
XM_021467155.1 | XP_021322830.1 | 4 - 5 | 332 | 119 | ||
GenBank mRNA | BC090299.1 | - | 4 - 5 | 401 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |