Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: napga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013480.2 | NP_001013498.2 | 3 - 4 | 293 | 66 | |
XM_017353827.2 | XP_017209316.1 | 3 - 4 | 247 | 66 | ||
GenBank mRNA | BC091448.1 | - | 3 - 4 | 295 | 66 | |
BC164747.1 | - | 3 - 4 | 236 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |