Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mybpc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013511.2 | NP_001013529.2 | 19 - 20 | 1953 | 102 | |
XM_017353828.2 | XP_017209317.1 | 18 - 19 | 2018 | 102 | ||
XM_021470159.1 | XP_021325834.1 | 19 - 20 | 1987 | 102 | ||
XM_021470160.1 | XP_021325835.1 | 18 - 19 | 2015 | 102 | ||
XM_021470161.1 | XP_021325836.1 | 18 - 19 | 1965 | 102 | ||
XM_021470162.1 | XP_021325837.1 | 18 - 19 | 1902 | 102 | ||
GenBank mRNA | BC091662.1 | - | 1947 | 102 | ||
EE327610.1 | - | 4 - 5 | 433 | 102 | ||
EE714901.1 | - | 3 - 4 | 311 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |