Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ca8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017571.2 | NP_001017571.2 | 3 - 4 | 800 | 127 | |
GenBank mRNA | BC092740.1 | - | 3 - 4 | 682 | 127 | |
BC164636.1 | - | 3 - 4 | 421 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |