Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhbdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017614.1 | NP_001017614.1 | 4 - 5 | 723 | 66 | |
XM_021481406.1 | XP_021337081.1 | 4 - 5 | 768 | 66 | ||
GenBank mRNA | BC092835.1 | - | 4 - 5 | 723 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |