Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: baiap2l1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017830.1 | NP_001017830.1 | 13 - 14 | 1612 | 57 | |
XM_009299879.3 | XP_009298154.1 | 13 - 14 | 1628 | 57 | ||
GenBank mRNA | BC092786.1 | - | 13 - 14 | 1612 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |