Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC092983.1 | - | 4 - 5 | 364 | 98 | |
BC152125.1 | - | 4 - 5 | 363 | 98 | ||
EH570638.1 | - | 1 - 2 | 334 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |