Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: capns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017899.2 | NP_001017899.2 | 8 - 9 | 612 | 107 | |
GenBank mRNA | BC092946.1 | - | 4 - 5 | 356 | 107 | |
CV490399.1 | - | 8 - 9 | 543 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |