Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020761.2 | NP_001018597.1 | 1 - 2 | 298 | 71 | |
GenBank mRNA | BC095668.1 | - | 1 - 2 | 300 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |