Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmbsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024388.1 | NP_001019559.1 | 3 - 4 | 283 | 94 | |
XM_005157569.4 | XP_005157626.1 | 3 - 4 | 266 | 94 | ||
GenBank mRNA | AB188810.1 | - | 3 - 4 | 241 | 94 | |
BC095373.1 | - | 3 - 4 | 283 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |