Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fdps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025471.1 | NP_001020642.1 | 9 - 10 | 945 | 111 | |
GenBank mRNA | BC097112.1 | - | 9 - 10 | 945 | 111 | |
BC164398.1 | - | 8 - 9 | 901 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |