Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320148.1 | NP_001307077.1 | 7 - 8 | 1141 | 73 | |
XM_017354091.2 | XP_017209580.1 | 7 - 8 | 1184 | 73 | ||
GenBank mRNA | BC146747.1 | - | 7 - 8 | 1137 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |