Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100403.1 | NP_001093873.1 | 8 - 9 | 3085 | 83 | |
GenBank mRNA | DQ017617.1 | - | 6 - 7 | 898 | 83 | |
EF567113.1 | - | 8 - 9 | 3085 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |