Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Gene Expression Assays
›
Search Tool
›
Search Results
› Dr03425516_g1
See other ANK3B GE Assays ›
Gene Symbol:
ank3b
Gene Name:
ankyrin 3b
Gene Aliases:
ank3, zgc:171763
Chromosome Location:
Chr.12: 7640825 - 7992115 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03425516_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ank3b
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_009306464.3
XP_009304739.1
40 - 41
10787
91
XM_009306467.3
XP_009304742.1
38 - 39
4960
91
XM_009306468.2
XP_009304743.1
37 - 38
4606
91
XM_009306469.3
XP_009304744.1
36 - 37
4876
91
XM_021480224.1
XP_021335899.1
43 - 44
10877
91
XM_021480225.1
XP_021335900.1
43 - 44
10877
91
XM_021480226.1
XP_021335901.1
42 - 43
10865
91
XM_021480227.1
XP_021335902.1
43 - 44
10862
91
XM_021480228.1
XP_021335903.1
42 - 43
10853
91
XM_021480229.1
XP_021335904.1
42 - 43
10850
91
XM_021480230.1
XP_021335905.1
42 - 43
10844
91
XM_021480231.1
XP_021335906.1
43 - 44
10850
91
XM_021480232.1
XP_021335907.1
42 - 43
10832
91
XM_021480233.1
XP_021335908.1
41 - 42
10820
91
XM_021480234.1
XP_021335909.1
42 - 43
10817
91
XM_021480235.1
XP_021335910.1
42 - 43
10817
91
XM_021480236.1
XP_021335911.1
43 - 44
10878
91
XM_021480237.1
XP_021335912.1
44 - 45
10676
91
XM_021480238.1
XP_021335913.1
41 - 42
10805
91
XM_021480239.1
XP_021335914.1
41 - 42
10799
91
XM_021480240.1
XP_021335915.1
43 - 44
10700
91
XM_021480241.1
XP_021335916.1
43 - 44
10877
91
XM_021480242.1
XP_021335917.1
42 - 43
10778
91
XM_021480243.1
XP_021335918.1
43 - 44
10837
91
XM_021480244.1
XP_021335919.1
43 - 44
10488
91
XM_021480245.1
XP_021335920.1
39 - 40
10426
91
XM_021480246.1
XP_021335921.1
39 - 40
10423
91
XM_021480247.1
XP_021335922.1
43 - 44
10878
91
XM_021480248.1
XP_021335923.1
39 - 40
10727
91
XM_021480249.1
XP_021335924.1
38 - 39
10673
91
XM_021480250.1
XP_021335925.1
43 - 44
10877
91
XM_021480251.1
XP_021335926.1
43 - 44
10877
91
XM_021480252.1
XP_021335927.1
43 - 44
10879
91
XM_021480253.1
XP_021335928.1
43 - 44
10879
91
XM_021480254.1
XP_021335929.1
20 - 21
8311
91
XM_021480255.1
XP_021335930.1
19 - 20
8299
91
XM_021480256.1
XP_021335931.1
18 - 19
8254
91
XM_021480257.1
XP_021335932.1
17 - 18
8220
91
XM_021480258.1
XP_021335933.1
42 - 43
5078
91
XM_021480259.1
XP_021335934.1
41 - 42
5051
91
XM_021480260.1
XP_021335935.1
40 - 41
5018
91
GenBank mRNA
BC151969.1
-
2 - 3
420
91
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
3' Most
Mammalian Gene Collection
Detect Genomic DNA
Danio rerio
Probe spans exons
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
signal transduction
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0