Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001819.2 | NP_001001819.1 | 3 - 4 | 347 | 106 | |
GenBank mRNA | AY561511.1 | - | 3 - 4 | 351 | 106 | |
BC076221.1 | - | 3 - 4 | 347 | 106 | ||
BC081516.1 | - | 3 - 4 | 347 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |