Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002139.1 | NP_001002139.1 | 5 - 6 | 524 | 60 | |
GenBank mRNA | AY648841.1 | - | 5 - 6 | 527 | 60 | |
BC071420.1 | - | 5 - 6 | 524 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |