Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapre3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002170.1 | NP_001002170.1 | 4 - 5 | 387 | 64 | |
XM_021474989.1 | XP_021330664.1 | 4 - 5 | 489 | 64 | ||
XM_021474990.1 | XP_021330665.1 | 3 - 4 | 400 | 64 | ||
XM_021474991.1 | XP_021330666.1 | 3 - 4 | 397 | 64 | ||
GenBank mRNA | BC074053.1 | - | 4 - 5 | 387 | 64 | |
BC164598.1 | - | 2 - 3 | 302 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |