Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002334.1 | NP_001002334.1 | 1 - 2 | 146 | 62 | |
GenBank mRNA | BC076516.1 | - | 1 - 2 | 146 | 62 | |
BC165139.1 | - | 1 - 2 | 99 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |