Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: etnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034175.1 | NP_001029347.1 | 7 - 8 | 1197 | 80 | |
GenBank mRNA | BC096922.1 | - | 7 - 8 | 1197 | 80 | |
BC164400.1 | - | 7 - 8 | 1038 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |