Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034978.2 | NP_001030150.2 | 5 - 6 | 1153 | 91 | |
GenBank mRNA | BC100113.1 | - | 5 - 6 | 1093 | 91 | |
EH552235.1 | - | 5 - 6 | 837 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |