Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drgx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037105.1 | NP_001032182.1 | 1 - 2 | 52 | 69 | |
XM_005156850.4 | XP_005156907.1 | 2 - 3 | 650 | 69 | ||
GenBank mRNA | AY753211.1 | - | 1 - 2 | 52 | 69 | |
BC162787.1 | - | 1 - 2 | 82 | 69 | ||
BC162791.1 | - | 1 - 2 | 82 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |