Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037243.2 | NP_001032320.2 | 4 - 5 | 1215 | 58 | |
XM_017353139.2 | XP_017208628.1 | 5 - 6 | 1093 | 58 | ||
XM_021469324.1 | XP_021324999.1 | 4 - 5 | 1226 | 58 | ||
GenBank mRNA | AB188254.1 | - | 4 - 5 | 1187 | 58 | |
BC163023.1 | - | 4 - 5 | 1180 | 58 | ||
BC163060.1 | - | 4 - 5 | 1180 | 58 | ||
BC163073.1 | - | 4 - 5 | 1180 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |