Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: golph3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037699.1 | NP_001032788.1 | 2 - 3 | 290 | 61 | |
GenBank mRNA | BC107999.1 | - | 2 - 3 | 290 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |