Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgals3bp.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040041.1 | NP_001035130.1 | 3 - 4 | 436 | 85 | |
GenBank mRNA | BC114268.1 | - | 3 - 4 | 436 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |