Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076558.1 | NP_001070026.1 | 4 - 5 | 538 | 95 | |
XM_005171872.3 | XP_005171929.2 | 4 - 5 | 495 | 95 | ||
XM_005171873.3 | XP_005171930.1 | 4 - 5 | 533 | 95 | ||
XM_005171874.4 | XP_005171931.1 | 4 - 5 | 519 | 95 | ||
GenBank mRNA | BC124351.1 | - | 4 - 5 | 538 | 95 | |
BC155131.1 | - | 4 - 5 | 419 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |