Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxred2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076660.1 | NP_001070128.1 | 8 - 9 | 1795 | 107 | |
XM_005163913.4 | XP_005163970.1 | 8 - 9 | 1824 | 107 | ||
XM_009299432.3 | XP_009297707.1 | 8 - 9 | 1938 | 107 | ||
GenBank mRNA | BC124275.1 | - | 8 - 9 | 1795 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |