Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oxct1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077150.1 | NP_001070618.1 | 14 - 15 | 1412 | 135 | |
GenBank mRNA | BC124298.1 | - | 14 - 15 | 1412 | 135 | |
BC154254.1 | - | 14 - 15 | 135 | |||
BC155216.1 | - | 14 - 15 | 135 | |||
BC164688.1 | - | 14 - 15 | 1396 | 135 |
Gene Symbol: LOC797311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021469231.1 | XP_021324906.1 | 12 - 13 | 1080 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |