Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgcra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079977.2 | NP_001073446.2 | 16 - 17 | 2350 | 64 | |
XM_005165515.4 | XP_005165572.1 | 16 - 17 | 2325 | 64 | ||
GenBank mRNA | BC117622.1 | - | 16 - 17 | 2354 | 64 | |
BC155135.1 | - | 16 - 17 | 2325 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |