Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tp53inp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080036.1 | NP_001073505.1 | 2 - 3 | 508 | 82 | |
GenBank mRNA | BC127596.1 | - | 2 - 3 | 508 | 82 | |
BC153523.1 | - | 1 - 2 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |