Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snapc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080163.2 | NP_001073632.1 | 3 - 4 | 245 | 73 | |
XM_005159085.4 | XP_005159142.1 | 3 - 4 | 543 | 73 | ||
GenBank mRNA | BC133151.1 | - | 3 - 4 | 245 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |