Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp3ccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080594.2 | NP_001074063.2 | 13 - 14 | 1658 | 68 | |
XM_005155313.4 | XP_005155370.1 | 13 - 14 | 1704 | 68 | ||
GenBank mRNA | BC129382.1 | - | 13 - 14 | 1649 | 68 | |
BC165627.1 | - | 13 - 14 | 1379 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |