Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp3ccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080594.2 | NP_001074063.2 | 4 - 5 | 803 | 80 | |
XM_005155313.4 | XP_005155370.1 | 4 - 5 | 834 | 80 | ||
XM_005155314.4 | XP_005155371.1 | 4 - 5 | 834 | 80 | ||
GenBank mRNA | BC129382.1 | - | 4 - 5 | 794 | 80 | |
BC165627.1 | - | 4 - 5 | 524 | 80 | ||
DN898177.1 | - | 3 - 4 | 278 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |