Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp3ccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080594.2 | NP_001074063.2 | 9 - 10 | 1385 | 99 | |
XM_005155313.4 | XP_005155370.1 | 9 - 10 | 1416 | 99 | ||
XM_005155314.4 | XP_005155371.1 | 9 - 10 | 1416 | 99 | ||
GenBank mRNA | BC129382.1 | - | 9 - 10 | 1376 | 99 | |
BC165627.1 | - | 9 - 10 | 1106 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |