Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loxl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080626.1 | NP_001074095.1 | 9 - 10 | 1705 | 58 | |
XM_009301553.3 | XP_009299828.1 | 8 - 9 | 3068 | 58 | ||
GenBank mRNA | BC129226.1 | - | 9 - 10 | 1705 | 58 | |
BC155756.1 | - | 9 - 10 | 1736 | 58 | ||
EF030482.1 | - | 8 - 9 | 1608 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |