Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loxl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080626.1 | NP_001074095.1 | 11 - 12 | 2058 | 75 | |
XM_009301553.3 | XP_009299828.1 | 10 - 11 | 3421 | 75 | ||
GenBank mRNA | BC129226.1 | - | 11 - 12 | 2058 | 75 | |
BC155756.1 | - | 11 - 12 | 2089 | 75 | ||
EF030482.1 | - | 10 - 11 | 1961 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |