Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anpepb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089325.1 | NP_001082794.1 | 8 - 9 | 1514 | 87 | |
GenBank mRNA | BC115260.1 | - | 8 - 9 | 1514 | 87 | |
BC154244.1 | - | 8 - 9 | 1515 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |