Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slco1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348086.1 | NP_001335015.1 | 1 - 2 | 49 | 73 | |
XM_021474933.1 | XP_021330608.1 | 2 - 3 | 67 | 73 | ||
GenBank mRNA | BC139695.1 | - | 1 - 2 | 76 | 73 | |
KP294314.1 | - | 1 - 2 | 40 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |