Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhcga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089577.1 | NP_001083046.1 | 3 - 4 | 596 | 131 | |
GenBank mRNA | AB286867.1 | - | 3 - 4 | 621 | 131 | |
BC139524.1 | - | 3 - 4 | 596 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |