Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: valopb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110280.1 | NP_001103750.1 | 4 - 5 | 1003 | 88 | |
GenBank mRNA | AY996588.1 | - | 4 - 5 | 1003 | 88 | |
KT008403.1 | - | 4 - 5 | 921 | 88 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Danio rerio Probe spans exons |