Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il13ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110837.1 | NP_001104307.1 | 2 - 3 | 379 | 89 | |
GenBank mRNA | BC162934.1 | - | 2 - 3 | 382 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |