Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111150.2 | NP_001104620.2 | 5 - 6 | 784 | 70 | |
GenBank mRNA | BC078263.1 | - | 5 - 6 | 777 | 70 | |
BC134022.1 | - | 5 - 6 | 779 | 70 | ||
BC155256.1 | - | 5 - 6 | 765 | 70 | ||
BC155771.1 | - | 5 - 6 | 762 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |