Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: opn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111164.1 | NP_001104634.1 | 3 - 4 | 905 | 81 | |
GenBank mRNA | EF043381.1 | - | 3 - 4 | 905 | 81 | |
KT008413.1 | - | 3 - 4 | 899 | 81 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Danio rerio Probe spans exons |