Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp4cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111168.1 | NP_001104638.1 | 6 - 7 | 731 | 57 | |
XM_009306399.3 | XP_009304674.1 | 6 - 7 | 741 | 57 | ||
GenBank mRNA | BC155609.1 | - | 6 - 7 | 731 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |