Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112733.1 | NP_001106204.1 | 26 - 27 | 3801 | 128 | |
GenBank mRNA | AF114427.2 | - | 26 - 27 | 3807 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |