Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp27c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113337.3 | NP_001106808.2 | 2 - 3 | 616 | 106 | |
GenBank mRNA | BC154632.1 | - | 2 - 3 | 525 | 106 | |
BC171576.1 | - | 2 - 3 | 301 | 106 | ||
BC171578.1 | - | 2 - 3 | 301 | 106 | ||
EH591543.1 | - | 2 - 3 | 605 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |